Anti-FKBP59 Mouse Monoclonal Antibody [clone: KN382/EC1]

Lieferant: Enzo Life Sciences
ADISRA1400D ADISRA1400F
ENZOADISRA1400DEA 290 EUR
ENZOADISRA1400D ENZOADISRA1400F
Anti-FKBP59 Mouse Monoclonal Antibody [clone: KN382/EC1]
Antikörper
FKBP59, also known as Hsp56/p59 is an immunophilin that is a component of Hsp70/Hsp90/steroid receptor complexes. Expression of FKBP59 is increased in response to heat shock and ischemic stress, and is thought to play a role in the folding and cytoplasmic/nuclear distribution of steroid receptors.

Type: Primary
Antigen: FKBP59
Clonality: Monoclonal
Clone: KN382/EC1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Monkey, Bovine, Hamster, Mink, Rabbit, Guinea Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR