Anti-SAP97 Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX210280R100
ENZOALX210280R100EA 698 EUR
ENZOALX210280R100
Anti-SAP97 Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes mouse and rat SAP97. Detects a band of ~140kDa by Western blot.

Type: Primary
Antigen: SAP97
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR