Anti-ETRA Sheep Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX210507AC250
ENZOALX210507AC250EA 586 EUR
ENZOALX210507AC250
Anti-ETRA Sheep Polyclonal Antibody
Antikörper
Endothelins are a family of peptides that are potent vasoconstrictors produced by mammalian vascular endothelial cells. The receptors for the endothelins are found on various tissues including, but not restricted to, lung and intestine.

  • For Immunohistochemistry, Western Blot

Recognizes an epitope on the cytoplasmic side of human, mouse and rat ETA-receptor. Detects bands of 34-52kDa. Does not block the binding of endothelins to the receptor.

Type: Primary
Antigen: ETRA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR