Anti-NNOS Rabbit Polyclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX210529C100
ENZOALX210529C100EA 665 EUR
ENZOALX210529C100
Anti-NNOS Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit
  • For Western Blot
Recognizes human nNOS. Detects a band of ~140kDa by Western blot.
Type: Primary
Antigen: NNOS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human