Anti-ITGAV Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX210537R100
ENZOALX210537R100EA 399 EUR
ENZOALX210537R100
Anti-ITGAV Rabbit Polyclonal Antibody
Antikörper
CD51 (integrin αV subunit) associates with CD61 (integrin β3 subunit) binding to the RGD sequence in extracellular matrix proteins like vitronectin, fibrinogen, thrombospondin, von Willebrand factor and osteopontin. The αV/β3 heterodimer is expressed in endothelial cells, leukocytes, NK cells, macrophages, neutrophils, platelets and several malignant tumor cells and seems to play a role in embryo implantation, angiogenesis, wound healing and cancer. αV/β3  is also highly expressed in osteoclasts being important for bone resorption.

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes cytoplasmic domain of CD51 of all species.

Type: Primary
Antigen: ITGAV
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR