Anti-ADIPOR1 Goat Polyclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX210645C200
ENZOALX210645C200EA 648 EUR
ENZOALX210645C200
Anti-ADIPOR1 Goat Polyclonal Antibody
Antikörper
Host: Goat
  • For Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes human, mouse and rat AdipoR1.
Type: Primary
Antigen: ADIPOR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human