Anti-MMP2 Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX210753R500
ENZOALX210753R500EA 208 EUR
ENZOALX210753R500
Anti-MMP2 Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit

  • For Immunoprecipitation, Western Blot

Recognizes human and marmoset MMP-2. Moderately cross-reacts with MMP-1. Does not cross-react with MMP-3, MMP-8 or MMP-9.

Type: Primary
Antigen: MMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR