Anti-MRP1 Mouse Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX801013C250 ALX801013C500
ENZOALX801013C250EA 706 EUR
ENZOALX801013C250 ENZOALX801013C500
Anti-MRP1 Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes the internal epitope (P1511SDLLQQRGL1520) of human MRP1. Does not cross-react with human P-glycoprotein, MDR3 P-glycoprotein or human MRP2 (cMOAT) to MRP5.

Type: Primary
Antigen: MRP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR