Anti-Nicotinic Acetylcholine Receptor Mouse Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX802002R100
ENZOALX802002R100EA 652 EUR
ENZOALX802002R100
Anti-Nicotinic Acetylcholine Receptor Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes the intracellular domain of skeletal muscle nAChR gamma and delta subunits in Torpedo californica and the delta subunit in mouse, rat, chicken, frog and fish. Detects bands of ~60kDa (gamma subunit ) and ~65kDa (delta subunit ) by Western blot. Under non-reducing conditions detects a band of ~130kDa (dimer of delta subunit). Does not cross-react with the alpha1 and beta1 subunits in muscle or the neuronal nAChRs.

Type: Primary
Antigen: NICOTINIC ACETYLCHOLINE RECEPTOR
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR