Anti-CANX Mouse Monoclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX804014R100
ENZOALX804014R100EA 890 EUR
ENZOALX804014R100
Anti-CANX Mouse Monoclonal Antibody
Antikörper
Calnexin, also referred to as IP90 or p88, is a ~88kDa chaperone protein found in the endoplastmic reticulum. It binds with partially folded immunoglobulin proteins and retains them in the endoplastmic reticulum until they are completely folded.
  • For Immunocytochemistry, Immunoprecipitation, Western Blot
Recognizes human and mouse calnexin. Detects a band of ~90kDa by Western blot.
Type: Primary
Antigen: CANX
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human