Anti-Enhancer for Ligands Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX804034C050
ENZOALX804034C050EA 414 EUR
ENZOALX804034C050
Anti-Enhancer for Ligands Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Enhancer for Ligands
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR