Anti-PMCA Mouse Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX804049R100
ENZOALX804049R100EA 784 EUR
ENZOALX804049R100
Anti-PMCA Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG2a

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes all known (currently four) isoforms of human, rat, mouse, sheep, dog, monkey, rabbit, cat, chicken and amphibian plasma membrane PMCA ATPase. Detects a band of ~140kDa by Western blot. Also detects bands at ~95kDa and ~180kDa which may represent aggregation and/or natural proteolytic products of PMCA ATPase. Recognizes an epitope between aa 724-783 of human PMCA ATPase.

Type: Primary
Antigen: PMCA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR