Anti-S Mouse Monoclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX804163C100
ENZOALX804163C100EA 535 EUR
ENZOALX804163C100
Anti-S Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgM
Recognizes a broad range of serine-phosphorylated proteins in crude cell extracts, interacting with phosphoserine surrounded by positive or neutral aa (e.g. substrates of PKA, PKB, PKC, PKG etc.).Please note that phosphoserine detection by MAbs always depends on the surrounding aa sequence.
For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Reconstitute with 1ml water (15 minutes at room temperature).
Bestellinformation: Positive control included.
Type: Primary
Antigen: S
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: pan