Anti-Ceramide Mouse Monoclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX804196T050
ENZOALX804196T050EA 730 EUR
ENZOALX804196T050
Anti-Ceramide Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgM
Recognizes C16- and C24-ceramide, dihydroceramide, sphingomyelin and phosphatidylcholine in highly artificial lipid overlay test systems. Under more physiological in vitro and in vivo conditions highly specific for ceramide and does not cross-react with sphingomyelin, cholesterol or other phospholipids.
For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Bestellinformation: Quantity: 1ml (50 tests). Suggested amount: 20µl/test.
Type: Primary
Antigen: Ceramide
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestellen