Anti-SHC1 Mouse Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX804358C100
ENZOALX804358C100EA 743 EUR
ENZOALX804358C100
Anti-SHC1 Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1

Recognizes human, mouse and dog shc phosphorylated at Ser36. Does not cross-react with non-phosphorylated shc.

For Western Blot

Reconstitute with 1ml water (15 minutes at room temperature).

Bestellinformation: Positive control included.

Type: Primary
Antigen: SHC1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR