Anti-CD14 Mouse Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX804496C100
ENZOALX804496C100EA 439 EUR
ENZOALX804496C100
Anti-CD14 Mouse Monoclonal Antibody
Antikörper
The CD14 glycoprotein, gp 55, is present on most monocytic and macrophage like cell types: monocytes, macrophages, Kupffer cells, pleural phagocytic cells and dendritic reticular cells. CD14 is also observed on granulocytes and activated or transformed B-cells. Furthermore CD14 is present in a soluble form in human serum, urine and other body fluids. The CD14 molecule has been reported to be a receptor for endotoxin.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human (epitope aa 9-13 & aa 39-44), pig, dog, horse and bovine CD14.

Reconstitute with 100µl distilled water.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR