Anti-IL13R Mouse Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX804506C100
ENZOALX804506C100EA 449 EUR
ENZOALX804506C100
Anti-IL13R Mouse Monoclonal Antibody
Antikörper
IL-13 is an immunoregulatory cytokine secreted predominantly by activated TH2 cells, as well as mast cells and basophils, and is a key mediator in the pathogenesis of allergic inflammation. The human IL-13 receptor (IL-13R) is a heterodimer composed of the interleukin-4 receptor a chain (IL-4Rα) and an IL-13 binding protein, IL-13Rα1.A second IL-13 binding protein, IL-13Rα2, has also been identified, which is believed to act as a "decoy" receptor. IL-13 receptors are expressed on human B cells, basophils, eosinophils, mast cells, endothelial cells, fibroblasts, monocytes, macrophages, respiratory epithelial cells, and smooth muscle cells. However, functional IL-13 receptors have not been demonstrated on human or mouse T cells.

Recognizes human IL-13R.

Type: Primary
Antigen: IL13R
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR