Anti-AKT Mouse Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ALX804553C100
ENZOALX804553C100EA 599 EUR
ENZOALX804553C100
Anti-AKT Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1

Recognizes a c-terminal epitope (aa 466-480) of phosphorylated and non-phosphorylated human, mouse, rat and dog Akt isoforms (see Technical Note below).

For ELISA, Flow Cytometry, Western Blot

Reconstitute with 1ml water (15 minutes at room temperature).

Bestellinformation: Positive control included.

Type: Primary
Antigen: AKT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR