Anti-TLR5 Mouse Monoclonal Antibody (R-PE) [clone: 85B152.5]

Lieferant: ENZO LIFE SCIENCES
ALX804597RC100
ENZOALX804597RC100EA 659 EUR
ENZOALX804597RC100
Anti-TLR5 Mouse Monoclonal Antibody (R-PE) [clone: 85B152.5]
Antikörper
Recommended Applications: Flow Cytometry

Species reactivity: Human, Mouse, Dog

Type: Primary
Antigen: TLR5
Clonality: Monoclonal
Clone: 85B152.5
Conjugation: R-PE
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR