Anti-LRRC32 Mouse Monoclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX804867C100
ENZOALX804867C100EA 664 EUR
ENZOALX804867C100
Anti-LRRC32 Mouse Monoclonal Antibody
Antikörper
LRRC32 (leucine rich repeat containing 32; also known as GARP or Garpin; Glycoprotein A repetitions predominant) is a glycoprotein expressed on the cell surface of megakaryocytes, platelets and activated regulatory T (Treg) cells.
  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Recognizes human and mouse LRRC32.
Type: Primary
Antigen: LRRC32
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human