Anti-ICAM1 Mouse Monoclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX805016C050
ENZOALX805016C050EA 429 EUR
ENZOALX805016C050
Anti-ICAM1 Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1
  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Recognizes the D1 domain of human CD54. Detects a band of ~90kDa by Western blot. Does not cross-react with mouse or rhesus monkey CD54.
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human