Anti-CD40L Mouse Monoclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ALX805037C100
ENZOALX805037C100EA 437 EUR
ENZOALX805037C100
Anti-CD40L Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1
Recognizes human CD40L.
Bestellinformation: Quantity: 100µl
Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen