Anti-CD41 Mouse Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
BMLIG60650100
ENZOBMLIG60650100EA 235 EUR
ENZOBMLIG60650100
Anti-CD41 Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Western Blot

Recognizes human alphaIIb integrin.

Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR