Fluor-de-Lys® Developer II concentrate
Lieferant: ENZO LIFE SCIENCES
FLUOR DE LYS® Developer II is an essential reagent for signal generation in assays done with the FLUOR DE LYS®-HDAC8, FLUOR DE LYS®-SIRT1 and FLUOR DE LYS®-SIRT2 substrates. Deacetylation of the FLUOR DE LYS® substrate sensitises it so that, in a second step, treatment with the FLUOR DE LYS® Developer II produces a fluorophore. Typically, an ‘inhibitor stop’ will be included in the 1X Developer dilution (e.g. nicotinamide for sirtuins).
Bestellinformation: Quantity: NOTE: This product is supplied packaged in 5 tubes (250 µl per tube) for a total volume of 1,25 ml. This packaging is used for optimal stability of the developer.