Anti-HERP Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
BMLPW97050025 BMLPW97050100
ENZOBMLPW97050025EA 157 EUR
ENZOBMLPW97050025 ENZOBMLPW97050100
Anti-HERP Rabbit Polyclonal Antibody
Antikörper
Homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domain member 1 protein (HERP) is a membrane-associated ER protein strongly up-regulated by inducers of ER stress.

  • For Immunoprecipitation, Western Blot

Recognizes human HERP.

Type: Primary
Antigen: HERP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR