Anti-COX-2 Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
BMLSA2370025 BMLSA2370100
ENZOBMLSA2370025EA 157 EUR
ENZOBMLSA2370025 ENZOBMLSA2370100
Anti-COX-2 Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and sheep COX-2. Does not cross-react with COX-1.

Type: Primary
Antigen: COX-2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR