Anti-Tubulin Alpha Rabbit Polyclonal Antibody
Lieferant: ENZO LIFE SCIENCES
BMLSA4520100
ENZOBMLSA4520100EA 533 EUR
ENZOBMLSA4520100
Anti-Tubulin Alpha Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit
  • For Western Blot
Recognizes primate, rodent, chicken, Xenopus and zebrafish alpha-tubulin. Detects a band of ~50kDa by Western blot.
Type: Primary
Antigen: TUBULIN ALPHA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Primate