Anti-SIRT5 Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
BMLSA4640100
ENZOBMLSA4640100EA 451 EUR
ENZOBMLSA4640100
Anti-SIRT5 Rabbit Polyclonal Antibody
Antikörper
SIRT5 is the only human class III sirtuin and as such is the closest homolog to bacterial (e.g. E. coli CobB) and archaeal sirtuins. SIRT5 is an imported mitochondrial protein with a calculated precursor molecular weight of 34kDa. In various human, rat, mouse and bovine cells and tissues, the major reactive band migrates with an apparent molecular weight of ~29kDa. This presumably reflects removal of an N-terminal transit sequence due to mitochondrial import and processing. No cross-reactivity observed with recombinant human sirtuins SIRT1, SIRT2 or SIRT3.

  • For Western Blot

Recognizes human, rat, mouse and bovine SIRT5. Detects a band of ~29 kDa by Western blot.

Type: Primary
Antigen: SIRT5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR