EpiXtract® total histone extraction kit (high throughput)
Lieferant: ENZO LIFE SCIENCES
ENZ450130002
ENZOENZ450130002EA 400 EUR
ENZOENZ450130002
EpiXtract® total histone extraction kit (high throughput)
Protein-Extraktion Protein Extraction Kits
Rapid, reliable, high throughput method to isolate histone proteins in mammalian cells for histone modification applications.
For the extraction of total histone proteins from mammalian cells in a 96 well plate format.
EpiXtract® Total Histone Extraction Kit (High Throughput) provides a simple and reliable method to isolate histone proteins in one hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays.
Bestellinformation: Contains Pre-Lysis Buffer, Lysis Buffer, Balance Buffer, DTT .