EpiXtract® total histone extraction kit
Lieferant: ENZO LIFE SCIENCES
Rapid, reliable method to isolate histone proteins in mammalian cells and tissues for histone modification applications.
For the extraction of total histone proteins from mammalian cells or tissues.
EpiXtract® Total Histone Extraction Kit provides a simple and reliable method to isolate histone proteins in one hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays.
Bestellinformation: Contains 10x Pre-Lysis Buffer, Lysis Buffer, Balance Buffer, DTT .