PROTEOSTAT® protein refolding and aggregation sensing kit for microplates
Lieferant: ENZO LIFE SCIENCES
ENZ51040KP002
ENZOENZ51040KP002EA 547 EUR
ENZOENZ51040KP002
PROTEOSTAT® protein refolding and aggregation sensing kit for microplates
Tests Proteintests
ProteoStat® Protein refolding and aggregation sensing kit provides a fractional factorial matrix design to facilitate screening of refolding parameters for a specific protein in different buffers, and a simple, homogenous assay format for monitoring protein aggregation using a red-emitting aggregation-sensitive dye. 
Bitte beachten RUO - Research Use Only.