Anti-NFH Mouse Monoclonal Antibody [clone: SMI-31]
Lieferant: ENZO LIFE SCIENCES
ENZABS2150100
ENZOENZABS2150100EA 707 EUR
ENZOENZABS2150100
Anti-NFH Mouse Monoclonal Antibody [clone: SMI-31]
Antikörper
Recommended Applications: ELISA, ICC, IHC, WB
Species reactivity: Human, Xenopus, Mammalian, Chicken
Type: Primary
Antigen: NFH
Clonality: Monoclonal
Clone: SMI-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human