Anti-NFH Mouse Monoclonal Antibody [clone: SMI-32]
Lieferant: ENZO LIFE SCIENCES
ENZABS2190100
ENZOENZABS2190100EA 707 EUR
ENZOENZABS2190100
Anti-NFH Mouse Monoclonal Antibody [clone: SMI-32]
Antikörper
Recommended Applications: ELISA, ICC, IHC, WB
Species reactivity: Mammalian
Type: Primary
Antigen: NFH
Clonality: Monoclonal
Clone: SMI-32
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human