Anti-CD23 Mouse Monoclonal Antibody [clone: 9p25]
Lieferant: ENZO LIFE SCIENCES
ENZABS2480100
ENZOENZABS2480100EA 578 EUR
ENZOENZABS2480100
Anti-CD23 Mouse Monoclonal Antibody [clone: 9p25]
Antikörper
Recommended Applications: Flow Cytometry, FUNC
Species reactivity: Human
Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: 9p25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human