Anti-Digoxigenin, Digoxin Sheep Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ENZABS2660100
ENZOENZABS2660100EA 238 EUR
ENZOENZABS2660100
Anti-Digoxigenin, Digoxin Sheep Polyclonal Antibody
Antikörper
Recommended Applications: ELISA, WB, TUNEL, ISH (in situ hybridization)

Species reactivity: Species independent

Type: Primary
Antigen: Digoxigenin, Digoxin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: E. coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR