Anti-Ebola virus Mouse Monoclonal Antibody [clone: FE25]

Lieferant: ENZO LIFE SCIENCES
ENZABS2670100
ENZOENZABS2670100EA 310 EUR
ENZOENZABS2670100
Anti-Ebola virus Mouse Monoclonal Antibody [clone: FE25]
Antikörper
Recommended Applications: ELISA, WB

Species reactivity: Ebola virus

Type: Primary
Antigen: Ebola virus
Clonality: Monoclonal
Clone: FE25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR