Anti-CHIP Rabbit Polyclonal Antibody
Lieferant: ENZO LIFE SCIENCES
ENZABS2730100
ENZOENZABS2730100EA 645 EUR
ENZOENZABS2730100
Anti-CHIP Rabbit Polyclonal Antibody
Antikörper
Recommended Applications: WB
Species reactivity: Human, Mouse
Type: Primary
Antigen: CHIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human