Pierce™ Digestion Indicator for Mass Spectrometry, Thermo Scientific
Lieferant: Thermo Fisher Scientific
The Digestion Indicator for Mass Spectrometry is a non-mammalian recombinant protein (26kDa) with five signature peptides for use in determining the digestion efficiency and reproducibility across multiple samples.
- Versatile
- For use with mass spectrometer
- Non-mammalian protein source
This product is sufficient for production of five signature peptides upon digestion for mass spectrometry.
Bitte beachten For Research Use Only. Not for use in diagnostic procedures.