Micrococcal nuclease solution (≥100 U/µl)
Lieferant: Thermo Fisher Scientific
88216
PIER88216EA
197
EUR
PIER88216
Micrococcal nuclease solution (≥100 U/µl)
Protein-Extraktion
Proteinextraktions-Reagenzien
This Micrococcal Nuclease (MNase) is suitable for removing nucleic acids from cell lysates, releasing chromatin-bound proteins and shearing chromatin for use in chromatin immunoprecipitation (ChIP) experiments.
- Enzyme digests nucleic acids (DNA and RNA)
- Effective for double-stranded, single-stranded, circular and linear nucleic acids
- Active in neutral to alkaline buffer conditions containing divalent calcium ions
- Highest activity is toward single-stranded nucleic acid substrates with preference for AT or AU rich regions