Protein quantitation reagent, TMTzero™ label reagent
Lieferant: Thermo Fisher Scientific
90067
PIER90067EA
229
EUR
PIER90067
Protein quantitation reagent, TMTzero™ label reagent
Protein-Extraktion
Proteinextraktions-Reagenzien
Pierce™ TMTzero™ Label Reagent serves as a "light" tag for MS-level peptide quantitation similar to duplex isotopic metabolic labeling, such as SILAC, or isotopic dimethylation labeling.