Anti-AMG 479 AL

Lieferant: ProSci Inc.
10-188
PRSI10-188EA 654 EUR
PRSI10-188
Anti-AMG 479 AL
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR