Anti-PF-04360365 Recombinant Antibody

Lieferant: ProSci Inc.
10-454
PRSI10-454EA 654 EUR
PRSI10-454
Anti-PF-04360365 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Fab-G1-kappa -[Fc-G1]
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR