Anti-VX-15 Recombinant Antibody

Lieferant: ProSci Inc.
10-695
PRSI10-695EA 654 EUR
PRSI10-695
Anti-VX-15 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa -[scFv]2
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR