Anti-GSK-2398852 Recombinant Antibody

Lieferant: ProSci Inc.
10-790
PRSI10-790EA 654 EUR
PRSI10-790
Anti-GSK-2398852 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: [VH-VH'-H-Gamma1-VL-VL'-C-kappa]-dimer
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR