Anti-MORAb-022 Recombinant Antibody

Lieferant: ProSci Inc.
10-846
PRSI10-846EA 654 EUR
PRSI10-846
Anti-MORAb-022 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2-kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR