Anti-XmAb-18087 Recombinant Antibody

Lieferant: ProSci Inc.
10-953
PRSI10-953EA 654 EUR
PRSI10-953
Anti-XmAb-18087 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa / scFv-h-CH2-CH3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR