Anti-MK-7684 Recombinant Antibody

Lieferant: ProSci Inc.
11-004
PRSI11-004EA 654 EUR
PRSI11-004
Anti-MK-7684 Recombinant Antibody
Antikörper
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR