Anti-ORF1a polyprotein SARS

Lieferant: ProSci Inc.
11-039
PRSI11-039EA 654 EUR
PRSI11-039
Anti-ORF1a polyprotein SARS
Antikörper
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR