Anti-MAP3K14 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
1129
PRSI1129EA 435 EUR
PRSI1129
Anti-MAP3K14 Rabbit Polyclonal Antibody
Antikörper
NIK Antibody: Nuclear factor kappa B (NF-kappa B) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa B mediates the expression of a great variety of genes in response to extracellular stimuli including IL-1, TNF alpha , LPS and mitogens. A serine/threonine protein kinase which mediates NF-kappa B activation by IL-1, TNF alpha and CD95 was identified recently and designated NIK (for NF-kappa B inducing kinase). NIK is an activator of I kappa B kinase alpha and beta (IKK alpha and IKK beta ). Therefore, NIK is a key molecule in the NF-kappa B signaling pathway leading to the induction of a variety of gene expression in response to proinflammatory cytokines and bacteria products.

Type:
Antigen: MAP3K14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR