Anti-CD300A Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
13-367
PRSI13-367EA 419 EUR
PRSI13-367
Anti-CD300A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD300A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR